missing translation for 'onlineSavingsMsg'
Learn More

Citalopram Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015042
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015042 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015042 Supplier Merck Emd Millipore Supplier No. PHR190150MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Citalopram Related Compound A is a process related impurity generated during the synthesis of Citalopram. Citalopram is a racemic bicyclic phthalane derivative and is a potent anti-depressant drug, that acts by inhibition of serotonin uptake.

Specifications

CAS 64372-56-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H23 FN2O2
Synonym 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarboxamide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 342.41
Grade Certified Reference Material