missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citalopram Hydrobromide 98.0+%, TCI America™
Supplier: TCI America C23705G
Specifications
| Citalopram Hydrobromide | |
| 183°C | |
| C20H22BrFN2O | |
| 5 g | |
| WIHMBLDNRMIGDW-UHFFFAOYSA-N | |
| 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide | |
| 77995 | |
| ≥98.0% (HPLC,T) |
| 59729-32-7 | |
| White | |
| MFCD02101306 | |
| citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan | |
| CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br | |
| 405.311 | |
| 405.31 | |
| Crystalline Powder |
Chemical Identifiers
| 59729-32-7 | |
| 405.311 | |
| WIHMBLDNRMIGDW-UHFFFAOYSA-N | |
| 77995 | |
| CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br |
| C20H22BrFN2O | |
| MFCD02101306 | |
| citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan | |
| 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide |
Safety and Handling
RTECSNumber : NP6406750
TSCA : No