missing translation for 'onlineSavingsMsg'
Learn More

Citalopram Hydrobromide 98.0+%, TCI America™

Catalog No. C23701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23701G 1 g
C23705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23701G Supplier TCI America Supplier No. C23701G

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H22BrFN2O
Molecular Weight (g/mol) 405.311
MDL Number MFCD02101306
InChI Key WIHMBLDNRMIGDW-UHFFFAOYSA-N
Synonym citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
PubChem CID 77995
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 405.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Citalopram Hydrobromide
RTECSNumber NP6406750
TSCA No