missing translation for 'onlineSavingsMsg'
Learn More

Citalopram Hydrobromide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014811
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014811 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014811 Supplier Merck Emd Millipore Supplier No. PHR16401G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 59729-32-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H21 FN2 O · HBr
MDL Number MFCD02101306
Synonym Citalopram hydrobromide; 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide
Type Certified Reference Material
Quantity 1 g
Formula Weight 405.30
Grade Certified Reference Material