missing translation for 'onlineSavingsMsg'
Learn More

cis-Stilbene 96.0+%, TCI America™

Catalog No. p-7135803
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00891ML 1 mL
S00895ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00891ML Supplier TCI America Supplier No. S00891ML
Only null left

Chemical Identifiers

CAS 645-49-8
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00004788
InChI Key PJANXHGTPQOBST-QXMHVHEDSA-N
Synonym cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
PubChem CID 5356785
ChEBI CHEBI:36008
IUPAC Name (Z)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 152°C
Quantity 1 mL
Formula Weight 180.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-Stilbene
RTECSNumber DA0513600
TSCA Yes