accessibility menu, dialog, popup

UserName

CAS RN 645-49-8

IUPAC Name: [(1Z)-2-phenylethenyl]benzene
Synonyms: cis-stilbene cis-diphenylethene isostilbene
Molecular Weight (g/mol): 180.25
Molecular Formula: C14H12
InChi Key: PJANXHGTPQOBST-QXMHVHEDSA-N
SMILES: C(=C\C1=CC=CC=C1)\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)