missing translation for 'onlineSavingsMsg'
Learn More

cis-Stilbene, 97%

Catalog No. p-7081776
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1192403 1 g
AAA1192406 5 g
AAA1192414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1192403 Supplier Thermo Scientific Chemicals Supplier No. A1192403
Only null left

CAS: 645-49-8 | C14H12 | 180.25 g/mol

cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

Solubility
Immiscible with water. Miscible with ethanol.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 645-49-8
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00004788
InChI Key PJANXHGTPQOBST-QXMHVHEDSA-N
Synonym cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
PubChem CID 5356785
ChEBI CHEBI:36008
IUPAC Name (Z)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 1°C to 2°C
Density 1.04
Boiling Point 307°C
Flash Point >110°C (230°F)
Odor Weak
Refractive Index 1.622
Quantity 1 g
Beilstein 1616739
Sensitivity Light sensitive
Merck Index 14,8817
Solubility Information Immiscible with water. Miscible with ethanol.
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material cis-Stilbene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
EINECSNumber 211-445-7
RTECSNumber DA0513600
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only