Learn More
cis-N-Boc-4-cyano-L-proline methyl ester, 97%
CAS: 487048-28-2 | C12H18N2O4 | 254.29 g/mol
Supplier: Thermo Scientific Chemicals H52783MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| cis-N-Boc-4-cyano-L-proline methyl ester | |
| C12H18N2O4 | |
| 250 mg | |
| GHLKJIQVORAVHE-BDAKNGLRSA-N | |
| 1-O-tert-butyl 2-O-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate | |
| 17750454 | |
| 97% |
| 487048-28-2 | |
| MFCD03787928 | |
| n-boc-cis-4-cyano-l-proline methyl ester, 2s,4s-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, boc-cis-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-cyanopyrrolidine-1,2-dicarboxylate, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, cis-n-boc-4-cyano-l-proline methyl ester 250mg, 2s,4s-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 2s,4s-4-cyano-1,2-pyrrolidinedicarboxylic acid 1-tert-butyl 2-methyl ester, 1,2-pyrrolidinedicarboxylic acid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4s | |
| COC(=O)[C@@H]1C[C@@H](CN1C(=O)OC(C)(C)C)C#N | |
| 254.29 | |
| 254.29 |
Chemical Identifiers
| 487048-28-2 | |
| 254.29 | |
| GHLKJIQVORAVHE-BDAKNGLRSA-N | |
| 17750454 | |
| COC(=O)[C@@H]1C[C@@H](CN1C(=O)OC(C)(C)C)C#N |
| C12H18N2O4 | |
| MFCD03787928 | |
| n-boc-cis-4-cyano-l-proline methyl ester, 2s,4s-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, boc-cis-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-cyanopyrrolidine-1,2-dicarboxylate, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4s, cis-n-boc-4-cyano-l-proline methyl ester 250mg, 2s,4s-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 2s,4s-4-cyano-1,2-pyrrolidinedicarboxylic acid 1-tert-butyl 2-methyl ester, 1,2-pyrrolidinedicarboxylic acid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4s | |
| 1-O-tert-butyl 2-O-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only