accessibility menu, dialog, popup

UserName

CAS RN 487048-28-2

CH3OO(S)(S)NOOH3CCH3H3CN

CAS RN 487048-28-2

IUPAC Name: 1-tert-butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 254.29
Molecular Formula: C12H18N2O4
InChi Key: GHLKJIQVORAVHE-BDAKNGLRSA-N
SMILES: COC(=O)[C@@H]1C[C@@H](CN1C(=O)OC(C)(C)C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results