Learn More
(+)-cis-Diltiazem hydrochloride
An L-type calcium channel blocker | CAS: 33286-22-5 | C22H27ClN2O4S | 450.978 g/mol
Supplier: Thermo Scientific Chemicals J6324506
Description
An L-type calcium channel blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (+)-cis-Diltiazem hydrochloride | |
| 207°C to 212°C | |
| C22H27ClN2O4S | |
| 5 g | |
| 14,3202 | |
| Soluble in water at 50mg/ml | |
| CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl | |
| 450.978 | |
| CHEBI:645509 | |
| Powder |
| 33286-22-5 | |
| White | |
| MFCD00069252 | |
| 4228706 | |
| diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem | |
| HDRXZJPWHTXQRI-BHDTVMLSSA-N | |
| [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride | |
| 62920 | |
| 450.98 |
Chemical Identifiers
| 33286-22-5 | |
| 450.978 | |
| HDRXZJPWHTXQRI-BHDTVMLSSA-N | |
| 62920 | |
| [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride |
| C22H27ClN2O4S | |
| MFCD00069252 | |
| diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem | |
| CHEBI:645509 | |
| CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 251-443-3
RTECSNumber : DL0310000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only