missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(triphenylphosphine)platinum(II), Premion™, 99.95% (metals basis), Pt 24.2% min

Catalog No. AA4100806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4100806 5 g
AA4100803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4100806 Supplier Thermo Scientific Chemicals Supplier No. 04100806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15604-36-1 | C36H30Cl2P2Pt | 790.57 g/mol

cis-Dichlorobis(triphenylphosphine)platinum(II) is used in hydrosilylation, wax oil catalytic cracking, platinum plating and as platinum catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15604-36-1
Molecular Formula C36H30Cl2P2Pt
Molecular Weight (g/mol) 790.57
MDL Number MFCD00010825
InChI Key ILIBUWCDNDIUOT-UHFFFAOYSA-L
Synonym cis-?dichlorobis triphenylphosphine ?platinum ii
PubChem CID 131667321
IUPAC Name platinum(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Pt++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 310°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Slightly soluble in chloroform,hexane and toluene.
Formula Weight 790.58
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 24.2% min.
Physical Form Powder
Assay (metals basis)
Chemical Name or Material cis-Dichlorobis(triphenylphosphine)platinum(II)
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only