missing translation for 'onlineSavingsMsg'
Learn More

cis-Diamminedichloroplatinum(II), Pt 64.5% min

Potent Anticancer Agent That Blocks DNA Synthesis. | CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol

$291.27 - $3816.59

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym Cisplatin
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1047101
View Documents
Thermo Scientific Chemicals
01047101
0.25 g
Each for $291.27
Only null left
 
AA1047103
View Documents
Thermo Scientific Chemicals
01047103
1 g
Each for $921.49
Only null left
 
AA1047106
View Documents
Thermo Scientific Chemicals
01047106
5 g
Each for $3,816.59
Only null left
 
Description

Description

Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15663-27-1
300.04
BSJGASKRWFKGMV-UHFFFAOYSA-L
dichloroplatinumbis(ylium) diamine
Cl2H6N2Pt
MFCD00011623
Cisplatin
N.N.Cl[Pt++]Cl

Specifications

270°C (decomposition)
Yellow
0.25 g
MFCD00011623
Soluble in DMF. Insoluble in most common solvents
N.N.Cl[Pt++]Cl
300.04
Odorless
cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis
15663-27-1
Micro-crystals
Cl2H6N2Pt
Cisplatin
BSJGASKRWFKGMV-UHFFFAOYSA-L
dichloroplatinumbis(ylium) diamine
300.06
Yellow micro crystals
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c

H300-H312+H332-H315-H318-H335-H350

DOTInformation : DOT Class: 6.1, Packing Group: II

EINECSNumber : 239-733-8

RTECSNumber : TP2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only