Learn More
cis-Diamminedichloroplatinum(II), Pt 64.5% min
Potent Anticancer Agent That Blocks DNA Synthesis. | CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol
Supplier: Thermo Scientific Chemicals 01047106
Description
Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| cis-Diamminedichloroplatinum(II) | |
| Potent anticancer agent that blocks DNA synthesis | |
| Yellow | |
| 5 g | |
| MFCD00011623 | |
| Soluble in DMF. Insoluble in most common solvents | |
| N.N.Cl[Pt++]Cl | |
| 300.04 | |
| Odorless |
| 270°C (decomposition) | |
| 15663-27-1 | |
| Micro-crystals | |
| Cl2H6N2Pt | |
| Cisplatin | |
| BSJGASKRWFKGMV-UHFFFAOYSA-L | |
| dichloroplatinumbis(ylium) diamine | |
| 300.06 | |
| Yellow micro crystals |
Chemical Identifiers
| 15663-27-1 | |
| 300.04 | |
| BSJGASKRWFKGMV-UHFFFAOYSA-L | |
| dichloroplatinumbis(ylium) diamine |
| Cl2H6N2Pt | |
| MFCD00011623 | |
| Cisplatin | |
| N.N.Cl[Pt++]Cl |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c
H300-H312+H332-H315-H318-H335-H350
DOTInformation : DOT Class: 6.1, Packing Group: II
EINECSNumber : 239-733-8
RTECSNumber : TP2450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only