missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Decahydronaphthalene 98.0+%, TCI America™
$121.16 - $855.60
Chemical Identifiers
| CAS | 493-01-6 |
|---|---|
| Molecular Formula | C10H18 |
| Molecular Weight (g/mol) | 138.25 |
| MDL Number | MFCD00004130,MFCD00064189,MFCD00064191 |
| InChI Key | NNBZCPXTIHJBJL-UHFFFAOYSA-N |
| Synonym | decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane |
| PubChem CID | 7044 |
| ChEBI | CHEBI:38853 |
| IUPAC Name | decahydronaphthalene |
| SMILES | C1CCC2CCCCC2C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D000925ML
|
TCI America
D000925ML |
25 mL |
Each for $121.16
|
|
|||||
|
D0009500ML
|
TCI America
D0009500ML |
500 mL |
Each for $855.60
|
|
|||||
Chemical Identifiers
| 493-01-6 | |
| 138.25 | |
| NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
| 7044 | |
| decahydronaphthalene |
| C10H18 | |
| MFCD00004130,MFCD00064189,MFCD00064191 | |
| decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
| CHEBI:38853 | |
| C1CCC2CCCCC2C1 |
Specifications
| 493-01-6 | |
| 193°C | |
| MFCD00004130,MFCD00064189,MFCD00064191 | |
| 1147 | |
| NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
| decahydronaphthalene | |
| 7044 | |
| 138.25 | |
| Liquid |
| Colorless | |
| C10H18 | |
| 25 mL | |
| decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
| C1CCC2CCCCC2C1 | |
| 138.25 | |
| CHEBI:38853 | |
| ≥98.0% (GC) | |
| cis-Decahydronaphthalene |
Safety and Handling
EINECSNumber : (4)-0575
RTECSNumber : QJ3150000
TSCA : Yes