missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-9,12-Octadecadienoyl-L-carnitine, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™

Catalog No. 111002136
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002136 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002136 Supplier Merck Emd Millipore Supplier No. 7677110MG

Specifications

CAS 36816-10-1
Linear Formula C25H45 NO4
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(9 Z,12 Z)-1-oxo-9,12-octadecadien-1-yl]oxy]-1-propanaminium inner salt; Linoleoyl-L-carnitine
Type Analytical Standard
Quantity 10 mg
Formula Weight 423.63
Percent Purity ≥95.0% (LC/MS)
Grade Analytical Standard