missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis,cis-9,12-Octadecadien-1-ol 95.0+%, TCI America™
$493.97 - $1297.38
Chemical Identifiers
| CAS | 506-43-4 |
|---|---|
| Molecular Formula | C18H34O |
| Molecular Weight (g/mol) | 266.47 |
| MDL Number | MFCD00056667 |
| InChI Key | JXNPEDYJTDQORS-HZJYTTRNSA-N |
| Synonym | Linoleyl Alcohol |
| PubChem CID | 5365682 |
| ChEBI | CHEBI:73534 |
| IUPAC Name | (9Z,12Z)-octadeca-9,12-dien-1-ol |
| SMILES | CCCCC\C=C/C\C=C/CCCCCCCCO |
Chemical Identifiers
| 506-43-4 | |
| 266.47 | |
| JXNPEDYJTDQORS-HZJYTTRNSA-N | |
| 5365682 | |
| (9Z,12Z)-octadeca-9,12-dien-1-ol |
| C18H34O | |
| MFCD00056667 | |
| Linoleyl Alcohol | |
| CHEBI:73534 | |
| CCCCC\C=C/C\C=C/CCCCCCCCO |
Specifications
| 506-43-4 | |
| 155°C | |
| MFCD00056667 | |
| Linoleyl Alcohol | |
| CCCCC\C=C/C\C=C/CCCCCCCCO | |
| 266.47 | |
| CHEBI:73534 | |
| ≥95.0% (GC) | |
| cis,cis-9,12-Octadecadien-1-ol |
| Yellow | |
| C18H34O | |
| 5 mL | |
| JXNPEDYJTDQORS-HZJYTTRNSA-N | |
| (9Z,12Z)-octadeca-9,12-dien-1-ol | |
| 5365682 | |
| 266.47 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0258
TSCA : Yes