missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-9,12-Octadecadien-1-ol 95.0+%, TCI America™

Supplier:  TCI America O022525ML

 View more versions of this product

Catalog No. O022525ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

cis,cis-9,12-Octadecadien-1-ol
Yellow
C18H34O
25 mL
JXNPEDYJTDQORS-HZJYTTRNSA-N
(9Z,12Z)-octadeca-9,12-dien-1-ol
5365682
266.47
Liquid
506-43-4
155°C
MFCD00056667
Linoleyl Alcohol
CCCCC\C=C/C\C=C/CCCCCCCCO
266.47
CHEBI:73534
≥95.0% (GC)

Chemical Identifiers

506-43-4
266.47
JXNPEDYJTDQORS-HZJYTTRNSA-N
5365682
(9Z,12Z)-octadeca-9,12-dien-1-ol
C18H34O
MFCD00056667
Linoleyl Alcohol
CHEBI:73534
CCCCC\C=C/C\C=C/CCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0258

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions