missing translation for 'onlineSavingsMsg'
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%

Catalog No. p-3593404
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367505000 500 mg Glass bottle
AC367500010 1 g Glass bottle
AC367500050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC367505000 Supplier Thermo Scientific Chemicals Supplier No. 367505000

CAS: 98014-14-3 | C20H16Cl2N4Ru | 484.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98014-14-3
Molecular Formula C20H16Cl2N4Ru
Molecular Weight (g/mol) 484.35
MDL Number MFCD00150005
InChI Key MGAJEYXQYJBLQL-UHFFFAOYSA-L
Synonym cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate, bis bipyridyl dichlororuthenium hydrate, 2,2'-bipyridine-dichlororuthenium-water 2/1/1, cis-dichlorobis 2,2'-bipyridine ruthenium ii, cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate, cis-bis 2,2-bipyridine dichlororuthenium ii hydrate, cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate, cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
PubChem CID 16211844
SMILES Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Based on Carbon)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: soluble
IUPAC Name dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
Formula Weight 484.35
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only