accessibility menu, dialog, popup

UserName

CAS RN 98014-14-3

IUPAC Name: bis(2,2'-bipyridine); dichlororuthenium
Synonyms: bis(bipyridyl); dichlororuthenium
Molecular Weight (g/mol): 484.35
Molecular Formula: C20H16Cl2N4Ru
InChi Key: MGAJEYXQYJBLQL-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results