missing translation for 'onlineSavingsMsg'
Learn More

cis-Aconitic anhydride, 98%

Catalog No. aaa1600606
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
AAA1600606 5 g
AAA1600603 1 g
AAA1600614 25 g
3 options

Catalog No. AAA1600606

Supplier: Thermo Scientific Chemicals A1600606

Only null left

CAS: 6318-55-4 | C6H4O5 | 156.093 g/mol

cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases. Store in a cool place.

Chemical Identifiers

CAS 6318-55-4
Molecular Formula C6H4O5
Molecular Weight (g/mol) 156.093
MDL Number MFCD00005524
InChI Key GVJRTUUUJYMTNQ-UHFFFAOYSA-N
Synonym cis-aconitic anhydride, aconitic anhydride, cis-aconitic acid anhydride, 3-furanacetic acid, 2,5-dihydro-2,5-dioxo, 2-2,5-dioxo-2,5-dihydrofuran-3-yl acetic acid, 2,5-dioxofuran-3-yl acetic acid, 2-2,5-dioxo-3-furyl acetic acid, 1-propene-1,2,3-tricarboxylic acid, cyclic 1,2-anhydride, 2,5-dioxo-2,5-dihydro-furan-3-yl-acetic acid, z-aconitic anhydride
PubChem CID 65163
IUPAC Name 2-(2,5-dioxofuran-3-yl)acetic acid
SMILES C1=C(C(=O)OC1=O)CC(=O)O

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
UN Number UN3261
Beilstein 127575
Sensitivity Moisture sensitive
Formula Weight 156.1
Percent Purity 98%
Chemical Name or Material cis-Aconitic anhydride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 228-663-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only