accessibility menu, dialog, popup

UserName

CAS RN 6318-55-4

IUPAC Name: 2-(2,5-dioxo-2,5-dihydrofuran-3-yl)acetic acid
Synonyms: (2,5-dioxofuran-3-yl)acetic acid
Molecular Weight (g/mol): 156.09
Molecular Formula: C6H4O5
InChi Key: GVJRTUUUJYMTNQ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=O)OC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results