missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Decene 98.0+%, TCI America™

Catalog No. D21631ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21631ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21631ML Supplier TCI America Supplier No. D21631ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7433-78-5
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00067488
InChI Key UURSXESKOOOTOV-KTKRTIGZSA-N
PubChem CID 5364449
IUPAC Name (Z)-dec-5-ene
SMILES CCCCC=CCCCC

Specifications

Color Colorless
Boiling Point 60°C
Quantity 1 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-5-Decene