accessibility menu, dialog, popup

UserName

CAS RN 7433-78-5

IUPAC Name: (5E)-dec-5-ene
Synonyms: trans-5-decene
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: UURSXESKOOOTOV-MDZDMXLPSA-N
SMILES: CCCC\C=C\CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results