missing translation for 'onlineSavingsMsg'
Learn More

cis-5,8,11,14,17-Eicosapentaenoic acid, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012360
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012360 100 mg
111012361 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012360 Supplier Merck Emd Millipore Supplier No. 44864100MG
Only null left

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 10417-94-4
Density 0.943 g/mL (at 25°C)
Flash Point 93°C
Physical Form Neat
Linear Formula CH3(CH2 CH=CH)5(CH2)3 CO2 H
MDL Number MFCD00065716
Melting Point -54°C to -53°C
Synonym EPA; Timnodonic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 302.45
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less