missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Octene 95.0+%, TCI America™

Catalog No. p-7134201
Change view
Click to view available options
Quantity:
1 mL
5 mL
Catalog No. Quantity
O01351ML 1 mL
O01355ML 5 mL
2 options

Catalog No. O01351ML

Supplier: TCI America O01351ML

Only null left

Chemical Identifiers

CAS 7642-15-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00067484
InChI Key IRUCBBFNLDIMIK-FPLPWBNLSA-N
Synonym cis-4-octene, z-oct-4-ene, z-4-octene, 4-octene, z, unii-19430whf5e, 4z-4-octene, 4z-oct-4-ene, 4-octene, 4z, cis-4-octene 1g, unii-68k818w2zc component
PubChem CID 5364446
IUPAC Name (4Z)-oct-4-ene
SMILES CCC\C=C/CCC

Specifications

Color Colorless
Boiling Point 122°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4-Octene
EINECSNumber (2)-0024
TSCA No