accessibility menu, dialog, popup

UserName

CAS RN 7642-15-1

IUPAC Name: (4Z)-oct-4-ene
Synonyms: (4Z)-oct-4-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: IRUCBBFNLDIMIK-FPLPWBNLSA-N
SMILES: CCC\C=C/CCC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)