missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Nonene 95.0+%, TCI America™

Catalog No. n03845ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03845ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N03845ML Supplier TCI America Supplier No. N03845ML
Only null left

Chemical Identifiers

CAS 10405-84-2
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00067487
InChI Key KPADFPAILITQBG-UHFFFAOYSA-N
PubChem CID 5364456
IUPAC Name non-4-ene
SMILES CCCCC=CCCC

Specifications

Color Colorless
Boiling Point 147°C
Quantity 5 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4-Nonene
EINECSNumber (2)-0025
TSCA No