accessibility menu, dialog, popup

UserName

CH3H3C(E/Z)

CAS RN 10405-84-2

IUPAC Name: non-4-ene
Synonyms: non-4-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: KPADFPAILITQBG-UHFFFAOYSA-N
SMILES: CCCCC=CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results