missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxycyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. H09805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09805G 5 g
H098025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09805G Supplier TCI America Supplier No. H09805G
Only null left

Chemical Identifiers

CAS 3685-22-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD04038017
InChI Key HCFRWBBJISAZNK-UHFFFAOYSA-N
PubChem CID 151138
IUPAC Name 4-hydroxycyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Hydroxycyclohexanecarboxylic Acid