CAS RN 3685-22-1
CAS RN 3685-22-1
IUPAC Name:
4-hydroxycyclohexane-1-carboxylic acid
Synonyms:
4-hydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
144.17
Molecular Formula:
C7H12O3
InChi Key:
HCFRWBBJISAZNK-UHFFFAOYSA-N
SMILES:
OC1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
cis-4-Hydroxycyclohexanecarboxylic Acid 98.0+%, TCI America™
CAS: 3685-22-1 Molecular Formula: C7H12O3 Molecular Weight (g/mol): 144.17 MDL Number: MFCD04038017 InChI Key: HCFRWBBJISAZNK-UHFFFAOYSA-N PubChem CID: 151138 IUPAC Name: 4-hydroxycyclohexane-1-carboxylic acid SMILES: C1CC(CCC1C(=O)O)O