missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Heptenal 95.0+%, TCI America™

Catalog No. p-7130579
Change view
Click to view available options
Quantity:
5 mL
25 mL
Catalog No. Quantity
H05205ML 5 mL
H052025ML 25 mL
2 options

Catalog No. H05205ML

Supplier: TCI America H05205ML

Only null left

Chemical Identifiers

CAS 6728-31-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00014682
InChI Key VVGOCOMZRGWHPI-ARJAWSKDSA-N
Synonym cis-4-heptenal, cis-4-hepten-1-al, z-hept-4-enal, z-4-heptenal, 4-heptenal, z, z-4-hepten-1-al, hept-cis-4-enal, unii-5bj99wwp64, 4-z-heptenal, hept-4 z-enal
PubChem CID 5362814
IUPAC Name (Z)-hept-4-enal
SMILES CCC=CCCC=O

Specifications

Color Yellow
Boiling Point 58°C
Quantity 5 mL
UN Number 1989
Formula Weight 112.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4-Heptenal
TSCA Yes
Recommended Storage Refrigerator