missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyclohexene-1,2-dicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. p-7124466
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C049325G 25 g
C0493500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C049325G Supplier TCI America Supplier No. C049325G
Only null left

Chemical Identifiers

CAS 935-79-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00005916
InChI Key KMOUUZVZFBCRAM-OLQVQODUSA-N
Synonym cis-1,2,3,6-tetrahydrophthalic anhydride, cis-4-cyclohexene-1,2-dicarboxylic anhydride, 3ar,7as-rel-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, unii-w9q4666nos, cis-3a,4,7,7a-tetrahydro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, cis, 3ar,7as-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, tetrahydrophthalic anhydride, cis, 3as,7ar-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione
PubChem CID 98495
IUPAC Name (3aS,7aR)-3a,4,7,7a-tetrahydro-2-benzofuran-1,3-dione
SMILES C1C=CCC2C1C(=O)OC2=O

Specifications

Melting Point 103°C
Color White
Boiling Point 195°C
Quantity 25 g
UN Number 2698
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic Anhydride
EINECSNumber (3)-3435
RTECSNumber GW5775000
TSCA Yes