accessibility menu, dialog, popup

UserName

CAS RN 935-79-5

IUPAC Name: (3aR,7aS)-1,3,3a,4,7,7a-hexahydro-2-benzofuran-1,3-dione
Synonyms: (3aR,7aS)-3a,4,7,7a-tetrahydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: KMOUUZVZFBCRAM-OLQVQODUSA-N
SMILES: O=C1OC(=O)[C@@H]2CC=CC[C@H]12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)