missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyclohexene-1,2-dicarboxylic anhydride, 95%

Catalog No. aab2143130
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2143130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2143130 Supplier Thermo Scientific Chemicals Supplier No. B2143130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 935-79-5 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935-79-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00005916
InChI Key KMOUUZVZFBCRAM-OLQVQODUSA-N
Synonym cis-1,2,3,6-tetrahydrophthalic anhydride, cis-4-cyclohexene-1,2-dicarboxylic anhydride, 3ar,7as-rel-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, unii-w9q4666nos, cis-3a,4,7,7a-tetrahydro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 1,3-isobenzofurandione, 3a,4,7,7a-tetrahydro-, cis, 3ar,7as-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione, tetrahydrophthalic anhydride, cis, 3as,7ar-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione
PubChem CID 98495
IUPAC Name (3aS,7aR)-3a,4,7,7a-tetrahydro-2-benzofuran-1,3-dione
SMILES C1C=CCC2C1C(=O)OC2=O

Specifications

Melting Point 97°C to 103°C
Quantity 250 g
Beilstein 82341
Sensitivity Moisture sensitive
Formula Weight 152.15
Percent Purity 95%
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic anhydride
Hazard Category H317-H318-H334-H335
Hazard Statement GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P271-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
EINECSNumber 213-308-7
RTECSNumber GW5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only