missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexylamine, 97%

Catalog No. aah6292514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6292514 25 g
AAH6292506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6292514 Supplier Thermo Scientific Chemicals Supplier No. H6292514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 247570-24-7 | C11H22N2O2 | 214.309 g/mol

cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexylamine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol, ethyl acetate and Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 247570-24-7
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD03844604
InChI Key FEYLUKDSKVSMSZ-UHFFFAOYSA-N
Synonym trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine
PubChem CID 2756050
IUPAC Name tert-butyl N-(4-aminocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)N

Specifications

Quantity 25 g
Solubility Information Soluble in methanol,ethyl acetate and Slightly soluble in water.
Formula Weight 214.3
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only