missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexanol, 97%

Catalog No. AAH6223806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6223806 5 g
AAH62238MD 250 mg
AAH6223803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6223806 Supplier Thermo Scientific Chemicals Supplier No. H6223806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 167081-25-6 | C11H21NO3 | 215.29 g/mol

cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

Solubility
Slightly soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 167081-25-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD03844613,MFCD03844614,MFCD06658349
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(O)CC1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water
Formula Weight 215.28
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only