CAS RN 167081-25-6
IUPAC Name:
tert-butyl N-(4-hydroxycyclohexyl)carbamate
Synonyms:
tert-butyl N-(4-hydroxycyclohexyl)carbamate
Molecular Weight (g/mol):
215.29
Molecular Formula:
C11H21NO3
InChi Key:
DQARDWKWPIRJEH-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1CCC(O)CC1
1
–
1
of
1
results
1
Promotions Available
cis-4-(Boc-amino)cyclohexanol, 97%
cis-4-(Boc-amino)cyclohexanol, 97%
CAS: 167081-25-6 Molecular Formula: C11H21NO3 Molecular Weight (g/mol): 215.29 MDL Number: MFCD03844613,MFCD03844614,MFCD06658349 InChI Key: DQARDWKWPIRJEH-UHFFFAOYSA-N Synonym: trans-4-boc-aminocyclohexanol,tert-butyl 4-hydroxycyclohexyl carbamate,boc-trans-4-aminocyclohexanol,4-boc-amino cyclohexanol,tert-butyl trans-4-hydroxycyclohexyl carbamate,tert-butyl n-4-hydroxycyclohexyl carbamate,tert-butyl cis-4-hydroxycyclohexylcarbamate,trans-n-boc-4-amino-cyclohexanol,4-n-boc-amino-cyclohexanol,tert-butyl cis-4-hydroxycyclohexyl carbamate PubChem CID: 1514287 IUPAC Name: tert-butyl N-(4-hydroxycyclohexyl)carbamate SMILES: CC(C)(C)OC(=O)NC1CCC(O)CC1