missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexanol, 97%

CAS: 167081-25-6 | C11H21NO3 | 215.29 g/mol

$98.86 - $1232.18

Chemical Identifiers

CAS 167081-25-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD03844613,MFCD03844614,MFCD06658349
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(O)CC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62238MD
View Documents
Thermo Scientific Chemicals
H62238MD
250 mg
Each for $98.86
Only null left
 
AAH6223803
View Documents
Thermo Scientific Chemicals
H6223803
1 g
Each for $287.74
Only null left
 
AAH6223806
View Documents
Thermo Scientific Chemicals
H6223806
5 g
Each for $1,232.18
Only null left
 
Description

Description

cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

Solubility
Slightly soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

167081-25-6
215.29
DQARDWKWPIRJEH-UHFFFAOYSA-N
1514287
CC(C)(C)OC(=O)NC1CCC(O)CC1
C11H21NO3
MFCD03844613,MFCD03844614,MFCD06658349
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
tert-butyl N-(4-hydroxycyclohexyl)carbamate

Specifications

167081-25-6
MFCD03844613,MFCD03844614,MFCD06658349
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
DQARDWKWPIRJEH-UHFFFAOYSA-N
tert-butyl N-(4-hydroxycyclohexyl)carbamate
1514287
97%
C11H21NO3
250 mg
Slightly soluble in water
CC(C)(C)OC(=O)NC1CCC(O)CC1
215.29
215.28
cis-4-(Boc-amino)cyclohexanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only