missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 6217-54-5 |
| Flash Point | 62°C |
| Physical Form | Neat |
| Linear Formula | C22H32O2 |
| MDL Number | MFCD00065722 |
| Synonym | DHA |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 328.49 |
| Percent Purity | ≥98.0% (GC) |
| Show More |