missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Octene 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 123°C |
| Quantity | 5 mL |
| UN Number | 3295 |
| Formula Weight | 112.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | cis-3-Octene |
Safety and Handling
| EINECSNumber | (2)-0024 |
| TSCA | No |