accessibility menu, dialog, popup

UserName

CH3CH3(Z)

CAS RN 14850-22-7

IUPAC Name: (3Z)-oct-3-ene
Synonyms: (3Z)-oct-3-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: YCTDZYMMFQCTEO-ALCCZGGFSA-N
SMILES: CCCC\C=C/CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results