missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Octen-1-ol, 95%

Catalog No. AAA1901409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1901409 10 g
AAA1901418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1901409 Supplier Thermo Scientific Chemicals Supplier No. A1901409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20125-84-2 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20125-84-2
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014062
InChI Key YDXQPTHHAPCTPP-WAYWQWQTSA-N
Synonym cis-3-octen-1-ol, z-3-octen-1-ol, z-oct-3-en-1-ol, 3-octen-1-ol, z, unii-34a1x2y8m9, 3z-octen-1-ol, 3z-3-octen-1-ol, 3-octenol, cis, fema no. 3467, 3-octen-1-ol, 3z
PubChem CID 5364519
IUPAC Name (Z)-oct-3-en-1-ol
SMILES CCCCC=CCCO

Specifications

Density 0.85
Boiling Point 88°C to 89°C (15 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.447
Quantity 10 g
Beilstein 1720075
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material cis-3-Octen-1-ol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 243-533-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only