accessibility menu, dialog, popup

UserName

CAS RN 20125-84-2

IUPAC Name: (3E)-oct-3-en-1-ol
Synonyms: (3E)-oct-3-en-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: YDXQPTHHAPCTPP-AATRIKPKSA-N
SMILES: CCCC\C=C\CCO
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)