missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexenyl Isobutyrate 95.0+%, TCI America™

Catalog No. B103525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B103525ML Supplier TCI America Supplier No. B103525ML
Only null left

Chemical Identifiers

CAS 41519-23-7
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00036529
InChI Key OSMAJVWUIUORGC-WAYWQWQTSA-N
Synonym Isobutyric Acid cis-3-Hexenyl Ester
PubChem CID 5352539
IUPAC Name [(Z)-hex-3-enyl] 2-methylpropanoate
SMILES CCC=CCCOC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexenyl Isobutyrate
EINECSNumber (2)-0760
RTECSNumber UA2470200
TSCA Yes