accessibility menu, dialog, popup

UserName

CAS RN 41519-23-7

CH3OOCH3CH3(Z)

CAS RN 41519-23-7

IUPAC Name: (3Z)-hex-3-en-1-yl 2-methylpropanoate
Synonyms: cis-3-hexenyl isobutyrate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: OSMAJVWUIUORGC-WAYWQWQTSA-N
SMILES: CC\C=C/CCOC(=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results