missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexene 97.0+%, TCI America™

Catalog No. H03985ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03985ML 5 mL
H03981ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03985ML Supplier TCI America Supplier No. H03985ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7642-09-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00085300
InChI Key ZQDPJFUHLCOCRG-WAYWQWQTSA-N
Synonym cis-3-hexene, z-3-hexene, 3-hexene, z, z-hex-3-ene, 3z-hex-3-ene, 3-hexene, 3z, unii-50181bz6yt, 3z-3-heptene, 3z-3-hexene #, 3-hexene cis
PubChem CID 643783
IUPAC Name (Z)-hex-3-ene
SMILES CCC=CCC

Specifications

Color Colorless
Boiling Point 67°C
Quantity 5 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexene
EINECSNumber (2)-0031
TSCA No