missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Hexen-1-ol 97.0+%, TCI America™
$100.62 - $258.21
Chemical Identifiers
| CAS | 928-96-1 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.16 |
| MDL Number | MFCD00063217 |
| InChI Key | UFLHIIWVXFIJGU-ARJAWSKDSA-N |
| Synonym | z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z |
| PubChem CID | 5281167 |
| ChEBI | CHEBI:28857 |
| IUPAC Name | (3Z)-hex-3-en-1-ol |
| SMILES | CC\C=C/CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H012425ML
|
N/A | 25 mL |
Each for $100.62
|
|
|||||
|
H0124100ML
|
N/A | 100 mL |
Each for $258.21
|
|
|||||
Chemical Identifiers
| C6H12O | |
| MFCD00063217 | |
| z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
| CHEBI:28857 | |
| CC\C=C/CCO |
Specifications
| 928-96-1 | |
| 157°C | |
| MFCD00063217 | |
| UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
| (3Z)-hex-3-en-1-ol | |
| 5281167 | |
| 100.16 | |
| Liquid |
| Yellow | |
| C6H12O | |
| z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
| CC\C=C/CCO | |
| 100.16 | |
| CHEBI:28857 | |
| ≥97.0% (GC) | |
| cis-3-Hexen-1-ol |
Safety and Handling
EINECSNumber : (2)-2393
RTECSNumber : MP8400000
TSCA : Yes