missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Hexen-1-ol 97.0+%, TCI America™
Supplier: TCI America H0124100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C6H12O | |
| MFCD00063217 | |
| z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
| CHEBI:28857 | |
| CC\C=C/CCO |
Specifications
| cis-3-Hexen-1-ol | |
| Yellow | |
| C6H12O | |
| MFCD00063217 | |
| UFLHIIWVXFIJGU-ARJAWSKDSA-N | |
| (3Z)-hex-3-en-1-ol | |
| 5281167 | |
| 100.16 | |
| Liquid |
| 928-96-1 | |
| 157°C | |
| 100 mL | |
| z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z | |
| CC\C=C/CCO | |
| 100.16 | |
| CHEBI:28857 | |
| ≥97.0% (GC) |
Safety and Handling
EINECSNumber : (2)-2393
RTECSNumber : MP8400000
TSCA : Yes