missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Chloroacrylic acid, 98+%

Catalog No. AAA1479803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1479803 1 g
AAA1479806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1479803 Supplier Thermo Scientific Chemicals Supplier No. A1479803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1609-93-4 | C3H3ClO2 | 106.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1609-93-4
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00004368
InChI Key MHMUCYJKZUZMNJ-UPHRSURJSA-N
Synonym cis-3-chloroacrylic acid, z-3-chloroacrylic acid, cis-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, z, cis-3-chloropropenoic acid, 2z-3-chloroacrylic acid, 2-propenoic acid, 3-chloro-, 2z, cis-.beta.-chloroacrylic acid, z-3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, z-8ci
PubChem CID 643794
ChEBI CHEBI:27397
IUPAC Name (Z)-3-chloroprop-2-enoic acid
SMILES OC(=O)\C=C/Cl

Specifications

Melting Point 60°C to 62°C
Quantity 1 g
UN Number UN3261
Beilstein 1720278
Formula Weight 106.51
Percent Purity ≥98%
Chemical Name or Material cis-3-Chloroacrylic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 216-545-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only