missing translation for 'onlineSavingsMsg'
Learn More

cis-3,3,5-Trimethylcyclohexanol 96.0+%, TCI America™

Catalog No. T087425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T087425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T087425G Supplier TCI America Supplier No. T087425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 933-48-2
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00070479
InChI Key BRRVXFOKWJKTGG-JGVFFNPUSA-N
PubChem CID 1550823
IUPAC Name (1R,5R)-3,3,5-trimethylcyclohexan-1-ol
SMILES CC1CC(CC(C1)(C)C)O

Specifications

Color White
Quantity 25 g
Formula Weight 142.24
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material cis-3,3,5-Trimethylcyclohexanol
EINECSNumber (3)-2294
RTECSNumber GW0876000
TSCA Yes